Assay Design
Gene Transcripts
- Gene Symbol
- LEAP2
- Entrez Gene ID
-
609750
- Gene Name
- liver enriched antimicrobial peptide 2
- Gene Aliases
-
-
- Chromosome Location
-
Chr.11: 21156051 - 21157197 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_847079.5 |
XP_852172.1 |
1 - 2 |
288 |
72 |
View Full Product Details