Assay Design
Gene Transcripts
- Gene Symbol
- ARL5C
- Entrez Gene ID
-
608600
- Gene Name
- ADP ribosylation factor like GTPase 5C
- Gene Aliases
-
-
- Chromosome Location
-
Chr.9: 23201466 - 23207640 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_846551.5 |
XP_851644.1 |
- |
445 |
63 |
View Full Product Details