Assay Design
Gene Transcripts
- Gene Symbol
- MED10
- Entrez Gene ID
-
607911
- Gene Name
- mediator complex subunit 10
- Gene Aliases
-
-
- Chromosome Location
-
Chr.34: 7327370 - 7334833 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_844767.3 |
XP_849860.2 |
- |
150 |
87 |
View Full Product Details