Assay Design
Gene Transcripts
- Gene Symbol
- PXMP4
- Entrez Gene ID
-
610008
- Gene Name
- peroxisomal membrane protein 4
- Gene Aliases
-
-
- Chromosome Location
-
Chr.24: 22916336 - 22930082 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_847386.5 |
XP_852479.2 |
2 - 3 |
249 |
66 |
View Full Product Details