Assay Design



Gene Transcripts

Gene Symbol
CDC42SE1
Entrez Gene ID
608900
Gene Name
CDC42 small effector 1
Gene Aliases
-
Chromosome Location
Chr.17: 60138497 - 60146344 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039043.1 NP_001034132.1 3 - 4 485 121
XM_014120563.2 XP_013976038.1 3 - 4 556 121
GenBank mRNA DN267318.1 - 3 - 4 485 121

View Full Product Details