Assay Design
Gene Transcripts
- Gene Symbol
- MIS18A
- Entrez Gene ID
-
609701
- Gene Name
- MIS18 kinetochore protein A
- Gene Aliases
-
-
- Chromosome Location
-
Chr.31: 27025657 - 27039001 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_847014.4 |
XP_852107.1 |
4 - 5 |
686 |
125 |
View Full Product Details