Assay Design



Gene Transcripts

Gene Symbol
CCM2
Entrez Gene ID
100856167
Gene Name
CCM2 scaffold protein
Gene Aliases
-
Chromosome Location
Chr.16: 10347801 - 10392560 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003639538.4 XP_003639586.2 - 1186 67
XM_005629571.3 XP_005629628.1 - 2003 67
XM_022403750.1 XP_022259458.1 - 1214 67
XM_022403751.1 XP_022259459.1 - 1189 67
XM_022403753.1 XP_022259461.1 - 1288 67
XM_022403754.1 XP_022259462.1 - 924 67

View Full Product Details