Assay Design



Gene Transcripts

Gene Symbol
CLEC3B
Entrez Gene ID
609596
Gene Name
C-type lectin domain family 3 member B
Gene Aliases
-
Chromosome Location
Chr.20: 43319786 - 43332161 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014121786.2 XP_013977261.1 1 - 2 275 81

View Full Product Details