Assay Design



Gene Transcripts

Gene Symbol
GUCA1A
Entrez Gene ID
609180
Gene Name
guanylate cyclase activator 1A
Gene Aliases
-
Chromosome Location
Chr.12: 10786284 - 10795974 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022426044.1 XP_022281752.1 2 - 3 2721 97

View Full Product Details