Assay Design



Gene Transcripts

Gene Symbol
TTC32
Entrez Gene ID
100856293
Gene Name
tetratricopeptide repeat domain 32
Gene Aliases
-
Chromosome Location
Chr.17: 14965950 - 14971227 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003639577.4 XP_003639625.1 2 - 3 470 143

View Full Product Details