Assay Design



Gene Transcripts

Gene Symbol
MANBAL
Entrez Gene ID
610351
Gene Name
mannosidase beta like
Gene Aliases
-
Chromosome Location
Chr.24: 25921827 - 25947363 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005635035.3 XP_005635092.1 3 - 4 314 74
XM_005635036.2 XP_005635093.1 2 - 3 335 74
XM_847833.5 XP_852926.1 2 - 3 347 74

View Full Product Details