Assay Design
Gene Transcripts
- Gene Symbol
- PCP4
- Entrez Gene ID
-
610723
- Gene Name
- Purkinje cell protein 4
- Gene Aliases
-
-
- Chromosome Location
-
Chr.31: 34529688 - 34584497 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_848266.3 |
XP_853359.1 |
2 - 3 |
198 |
83 |
View Full Product Details