Assay Design
Gene Transcripts
- Gene Symbol
- SPINK2
- Entrez Gene ID
-
611439
- Gene Name
- serine peptidase inhibitor, Kazal type 2
- Gene Aliases
-
-
- Chromosome Location
-
Chr.13: 48952869 - 48964862 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_849107.5 |
XP_854200.1 |
1 - 2 |
156 |
108 |
View Full Product Details