Assay Design



Gene Transcripts

Gene Symbol
MYADM
Entrez Gene ID
611521
Gene Name
myeloid associated differentiation marker
Gene Aliases
-
Chromosome Location
Chr.1: 103315606 - 103322329 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005616122.3 XP_005616179.1 2 - 3 299 91
XM_022427529.1 XP_022283237.1 2 - 3 105 91
XM_849198.4 XP_854291.1 2 - 3 185 91

Gene Symbol
LOC100855913
Entrez Gene ID
100855913
Gene Name
myeloid-associated differentiation marker
Gene Aliases
-
Chromosome Location
Chr.1: 103515509 - 103522232 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003638804.3 XP_003638852.1 2 - 3 185 91
XM_005616198.3 XP_005616255.1 2 - 3 299 91
XM_022422994.1 XP_022278702.1 2 - 3 105 91

View Full Product Details