Assay Design
Gene Transcripts
- Gene Symbol
- PARP16
- Entrez Gene ID
-
610816
- Gene Name
- poly(ADP-ribose) polymerase family member 16
- Gene Aliases
-
-
- Chromosome Location
-
Chr.30: 29626675 - 29649992 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_848370.5 |
XP_853463.2 |
1 - 2 |
367 |
61 |
View Full Product Details