Assay Design



Gene Transcripts

Gene Symbol
TP53I13
Entrez Gene ID
611028
Gene Name
tumor protein p53 inducible protein 13
Gene Aliases
-
Chromosome Location
Chr.9: 43671184 - 43676330 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022423399.1 XP_022279107.1 2 - 3 251 151
XM_022423400.1 XP_022279108.1 2 - 3 250 151
XM_848643.4 XP_853736.1 2 - 3 252 151

View Full Product Details