Assay Design



Gene Transcripts

Gene Symbol
CMPK1
Entrez Gene ID
610291
Gene Name
cytidine/uridine monophosphate kinase 1
Gene Aliases
CMPK
Chromosome Location
Chr.15: 13324235 - 13357387 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022427957.1 XP_022283665.1 2 - 3 232 68
XM_847756.6 XP_852849.3 2 - 3 342 68

View Full Product Details