Assay Design
Gene Transcripts
- Gene Symbol
- MRPL53
- Entrez Gene ID
-
612261
- Gene Name
- mitochondrial ribosomal protein L53
- Gene Aliases
-
-
- Chromosome Location
-
Chr.17: 48663271 - 48666078 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_850000.5 |
XP_855093.1 |
2 - 3 |
247 |
99 |
View Full Product Details