Assay Design



Gene Transcripts

Gene Symbol
PMVK
Entrez Gene ID
612251
Gene Name
phosphomevalonate kinase
Gene Aliases
-
Chromosome Location
Chr.7: 42532968 - 42563618 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014115379.2 XP_013970854.1 1 - 2 190 77
XM_014115381.2 XP_013970856.1 1 - 2 61 77
XM_014115382.2 XP_013970857.1 1 - 2 204 77
XM_849989.5 XP_855082.1 1 - 2 183 77

View Full Product Details