Assay Design



Gene Transcripts

Gene Symbol
GTF2H5
Entrez Gene ID
612182
Gene Name
general transcription factor IIH subunit 5
Gene Aliases
-
Chromosome Location
Chr.1: 47669196 - 47681370 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005615543.3 XP_005615600.1 - 226 87
XM_022418418.1 XP_022274126.1 - 121 87
XM_849919.5 XP_855012.1 - 188 87

View Full Product Details