Assay Design
Gene Transcripts
- Gene Symbol
- ZCCHC24
- Entrez Gene ID
-
610507
- Gene Name
- zinc finger CCHC-type containing 24
- Gene Aliases
-
-
- Chromosome Location
-
Chr.4: 29103393 - 29161626 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_848028.6 |
XP_853121.1 |
1 - 2 |
337 |
82 |
View Full Product Details