Assay Design
Gene Transcripts
- Gene Symbol
- PPM1D
- Entrez Gene ID
-
612934
- Gene Name
- protein phosphatase, Mg2+/Mn2+ dependent 1D
- Gene Aliases
-
-
- Chromosome Location
-
Chr.9: 35856277 - 35914096 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_847666.5 |
XP_852759.2 |
1 - 2 |
681 |
65 |
View Full Product Details