Assay Design



Gene Transcripts

Gene Symbol
PPM1D
Entrez Gene ID
612934
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1D
Gene Aliases
-
Chromosome Location
Chr.9: 35856277 - 35914096 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022423258.1 XP_022278966.1 3 - 4 1035 93
XM_847666.5 XP_852759.2 3 - 4 1036 93

View Full Product Details