Assay Design



Gene Transcripts

Gene Symbol
MGP
Entrez Gene ID
100856382
Gene Name
matrix Gla protein
Gene Aliases
-
Chromosome Location
Chr.27: 31735301 - 31738673 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003639976.4 XP_003640024.1 1 - 2 155 154

View Full Product Details