Assay Design
Gene Transcripts
- Gene Symbol
- PTS
- Entrez Gene ID
-
611326
- Gene Name
- 6-pyruvoyltetrahydropterin synthase
- Gene Aliases
-
-
- Chromosome Location
-
Chr.5: 20883128 - 20912318 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_848983.5 |
XP_854076.1 |
- |
349 |
112 |
View Full Product Details