Assay Design
Gene Transcripts
- Gene Symbol
- UBL4A
- Entrez Gene ID
-
612563
- Gene Name
- ubiquitin like 4A
- Gene Aliases
-
-
- Chromosome Location
-
Chr.X: 122195500 - 122196579 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_850295.6 |
XP_855388.3 |
3 - 4 |
444 |
69 |
View Full Product Details