Assay Design
Gene Transcripts
- Gene Symbol
- PGLYRP1
- Entrez Gene ID
-
612209
- Gene Name
- peptidoglycan recognition protein 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 109614404 - 109618242 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_849945.4 |
XP_855038.1 |
2 - 3 |
451 |
83 |
View Full Product Details