Assay Design



Gene Transcripts

Gene Symbol
PRPF18
Entrez Gene ID
477998
Gene Name
pre-mRNA processing factor 18
Gene Aliases
-
Chromosome Location
Chr.2: 22868184 - 22922647 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005617120.3 XP_005617177.1 - 396 73
XM_535182.6 XP_535182.2 - 368 73

View Full Product Details