Assay Design



Gene Transcripts

Gene Symbol
INHBE
Entrez Gene ID
100856447
Gene Name
inhibin subunit beta E
Gene Aliases
-
Chromosome Location
Chr.10: 1558876 - 1563370 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005625539.3 XP_005625596.1 - 818 69

View Full Product Details