Assay Design
Gene Transcripts
- Gene Symbol
- H3-3B
- Entrez Gene ID
-
475916
- Gene Name
- H3.3 histone B
- Gene Aliases
-
H3F3B
- Chromosome Location
-
Chr.9: 4805655 - 4808076 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_853711.4 |
XP_858804.1 |
1 - 1 |
287 |
92 |
View Full Product Details