Assay Design
Gene Transcripts
- Gene Symbol
- WDR82
- Entrez Gene ID
-
484740
- Gene Name
- WD repeat domain 82
- Gene Aliases
-
WDR82P1
- Chromosome Location
-
Chr.20: 37494633 - 37513746 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_854531.5 |
XP_859624.1 |
6 - 7 |
1010 |
84 |
View Full Product Details