Assay Design



Gene Transcripts

Gene Symbol
FXYD2
Entrez Gene ID
608594
Gene Name
FXYD domain containing ion transport regulator 2
Gene Aliases
-
Chromosome Location
Chr.5: 15812335 - 15824552 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252337.1 NP_001239266.1 - 251 83
XM_005619533.3 XP_005619590.1 - 796 83
XM_005619534.3 XP_005619591.2 - 319 83
XM_005619536.3 XP_005619593.1 - 801 83
XM_005619537.3 XP_005619594.2 - 225 83
XM_014113204.2 XP_013968679.1 - 289 83
XM_014113205.2 XP_013968680.1 - 389 83
GenBank mRNA DN330290.1 - - 251 83
DN348014.1 - - 150 83

View Full Product Details