Assay Design



Gene Transcripts

Gene Symbol
BRK1
Entrez Gene ID
607701
Gene Name
BRICK1 subunit of SCAR/WAVE actin nucleating complex
Gene Aliases
-
Chromosome Location
Chr.20: 8220109 - 8231337 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005632170.2 XP_005632227.2 - 195 80

View Full Product Details