Assay Design
Gene Transcripts
- Gene Symbol
- LSM10
- Entrez Gene ID
-
475335
- Gene Name
- LSM10, U7 small nuclear RNA associated
- Gene Aliases
-
-
- Chromosome Location
-
Chr.15: 5886029 - 5888859 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_532561.6 |
XP_532561.1 |
1 - 2 |
195 |
144 |
View Full Product Details