Assay Design



Gene Transcripts

Gene Symbol
GSG1
Entrez Gene ID
486677
Gene Name
germ cell associated 1
Gene Aliases
-
Chromosome Location
Chr.27: 33286460 - 33302080 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014108271.2 XP_013963746.1 5 - 5 992 89
XM_543804.6 XP_543804.2 5 - 5 871 89

View Full Product Details