Assay Design
Gene Transcripts
- Gene Symbol
- FAHD1
- Entrez Gene ID
-
490068
- Gene Name
- fumarylacetoacetate hydrolase domain containing 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.6: 39082536 - 39084248 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_547188.5 |
XP_547188.1 |
1 - 1 |
630 |
132 |
View Full Product Details