Assay Design
Gene Transcripts
- Gene Symbol
- PPM1L
- Entrez Gene ID
-
608708
- Gene Name
- protein phosphatase, Mg2+/Mn2+ dependent 1L
- Gene Aliases
-
-
- Chromosome Location
-
Chr.34: 26770442 - 27060892 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_845816.5 |
XP_850909.1 |
4 - 4 |
1019 |
107 |
View Full Product Details