Assay Design



Gene Transcripts

Gene Symbol
ZC4H2
Entrez Gene ID
491920
Gene Name
zinc finger C4H2-type containing
Gene Aliases
-
Chromosome Location
Chr.X: 49783962 - 49852098 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005641416.3 XP_005641473.1 2 - 3 409 92
XM_549040.6 XP_549040.3 2 - 3 406 92

View Full Product Details