Assay Design
Gene Transcripts
- Gene Symbol
- RASD1
- Entrez Gene ID
-
489542
- Gene Name
- ras related dexamethasone induced 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.5: 41965091 - 41966812 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_546661.6 |
XP_546661.2 |
2 - 2 |
611 |
94 |
View Full Product Details