Assay Design



Gene Transcripts

Gene Symbol
CHST9
Entrez Gene ID
100688744
Gene Name
carbohydrate sulfotransferase 9
Gene Aliases
-
Chromosome Location
Chr.7: 61612085 - 61806726 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022421508.1 XP_022277216.1 5 - 5 1043 116
XM_022421509.1 XP_022277217.1 4 - 4 1004 116
XM_022421510.1 XP_022277218.1 5 - 5 933 116

View Full Product Details