Assay Design



Gene Transcripts

Gene Symbol
SPIN4
Entrez Gene ID
480938
Gene Name
spindlin family member 4
Gene Aliases
-
Chromosome Location
Chr.X: 48805292 - 48809546 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005641408.3 XP_005641465.1 1 - 1 787 127

View Full Product Details