Assay Design
Gene Transcripts
- Gene Symbol
- SMAD6
- Entrez Gene ID
-
487612
- Gene Name
- SMAD family member 6
- Gene Aliases
-
-
- Chromosome Location
-
Chr.30: 30938908 - 31014259 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_544737.7 |
XP_544737.2 |
1 - 1 |
1331 |
52 |
View Full Product Details