Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005633484.3 | XP_005633541.1 | 2 - 3 | 1799 | 132 |
XM_022407667.1 | XP_022263375.1 | 3 - 4 | 385 | 132 | |
XM_022407668.1 | XP_022263376.1 | 4 - 5 | 441 | 132 | |
XM_022407669.1 | XP_022263377.1 | 4 - 5 | 635 | 132 | |
XM_022407670.1 | XP_022263378.1 | 4 - 5 | 445 | 132 | |
XM_022407671.1 | XP_022263379.1 | 3 - 4 | 361 | 132 | |
XM_022407672.1 | XP_022263380.1 | 4 - 5 | 395 | 132 | |
XM_022407673.1 | XP_022263381.1 | 3 - 4 | 467 | 132 | |
XM_022407674.1 | XP_022263382.1 | 2 - 3 | 336 | 132 | |
XM_534007.6 | XP_534007.3 | 2 - 3 | 1799 | 132 |