Assay Design



Gene Transcripts

Gene Symbol
VTCN1
Entrez Gene ID
483140
Gene Name
V-set domain containing T cell activation inhibitor 1
Gene Aliases
-
Chromosome Location
Chr.17: 54533153 - 54587262 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005630662.3 XP_005630719.1 3 - 3 528 140

View Full Product Details