Assay Design
Gene Transcripts
- Gene Symbol
- LRRTM2
- Entrez Gene ID
-
481529
- Gene Name
- leucine rich repeat transmembrane neuronal 2
- Gene Aliases
-
-
- Chromosome Location
-
Chr.11: 26392642 - 26397018 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_538650.6 |
XP_538650.2 |
2 - 2 |
2270 |
80 |
View Full Product Details