Assay Design
Gene Transcripts
- Gene Symbol
- SHH
- Entrez Gene ID
-
608860
- Gene Name
- sonic hedgehog signaling molecule
- Gene Aliases
-
-
- Chromosome Location
-
Chr.16: 18650553 - 18663571 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_845357.4 |
XP_850450.1 |
- |
922 |
138 |
View Full Product Details