Assay Design
Gene Transcripts
- Gene Symbol
- PEX12
- Entrez Gene ID
-
491139
- Gene Name
- peroxisomal biogenesis factor 12
- Gene Aliases
-
-
- Chromosome Location
-
Chr.9: 38083895 - 38088192 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_548259.6 |
XP_548259.1 |
1 - 2 |
1014 |
143 |
View Full Product Details