Assay Design



Gene Transcripts

Gene Symbol
BLVRB
Entrez Gene ID
476456
Gene Name
biliverdin reductase B
Gene Aliases
-
Chromosome Location
Chr.1: 113244484 - 113258170 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022425584.1 XP_022281292.1 3 - 3 460 147
XM_533666.6 XP_533666.2 4 - 4 545 147

View Full Product Details