Assay Design
Gene Transcripts
- Gene Symbol
- NCCRP1
- Entrez Gene ID
-
612546
- Gene Name
- NCCRP1, F-box associated domain containing
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 113989951 - 113993847 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_850276.4 |
XP_855369.1 |
1 - 1 |
335 |
148 |
View Full Product Details