Assay Design



Gene Transcripts

Gene Symbol
OR5C1
Entrez Gene ID
609809
Gene Name
olfactory receptor family 5 subfamily C member 1
Gene Aliases
-
Chromosome Location
Chr.9: 59984662 - 59985621 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005625448.2 XP_005625505.1 1 - 1 566 150

View Full Product Details